This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277857_m1
See other 5830432E09RIK GE Assays ›
Gene Symbol:
5830432E09Rik
Gene Name:
RIKEN cDNA 5830432E09 gene
Gene Aliases:
-
Chromosome Location:
Chr.7: 135648457 - 135652314 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277857_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5830432E09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015548.1 2 - 3 690 92
GenBank mRNA AK017962.1 - 2 - 3 691 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1962 Mir1962 mir-1962,
mmu-mir-1962
-82.2kb Anti-sense Chr. 7 - 135566162 - 135566282 on NCBI Build GRCm38
Downstream antigen identified by monoclonal antibody Ki 67 Mki67 D630048A14Rik,
Ki-67,
Ki67
38.8kb Sense Chr. 7 - 135689782 - 135716408 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004450.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2241663 AK148045.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P345298 BY548245.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P345298 NR_015548.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P345298 AK017962.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430799_PM_at - -
Mouse Genome 430 2.0 Array 1430799_at - -
Mouse Genome 430 2.0 Array 1430799_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700037379.mm.1 NR_015548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037379.mm.1 NR_015548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037379.mm.1 TC0700004450.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700037381.mm.1 NR_015548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037381.mm.1 NR_015548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037381.mm.1 TC0700004450.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700037378.mm.1 NR_015548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037378.mm.1 NR_015548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037378.mm.1 TC0700004450.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700037380.mm.1 NR_015548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037380.mm.1 NR_015548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037380.mm.1 TC0700004450.mm.1 MTA TC
Mouse Expression Set 430 1430799_PM_at - -
Mouse Expression Set 430 1430799_at - -
Affymetrix HT Mouse Genome 430B Array 1430799_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430799_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 683868 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4780717 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0