This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277571_m1
See other E130008D07RIK GE Assays ›
Gene Symbol:
E130008D07Rik
Gene Name:
RIKEN cDNA E130008D07 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 43146041 - 43158291 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277571_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E130008D07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045153.1 4 - 5 1983 76
NR_045154.1 4 - 5 881 76
NR_045155.1 4 - 5 869 76
GenBank mRNA AK053296.1 - 4 - 5 846 76
AK132380.1 - 4 - 5 883 76
BC072601.1 - 4 - 5 1888 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM949605.1 - 4 - 5 367 Chr. 17 - 43151964 - 43157845 [-] on Build GRCm38 76
GenBankmRNA BM949762.1 - 4 - 5 367 Chr. 17 - 43138397 - 43157845 [-] on Build GRCm38 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tumor necrosis factor receptor superfamily,
member 21
Tnfrsf21 AA959878,
DR6,
R74815,
TR7
-62.8kb Anti-sense Chr. 17 - 43016555 - 43089188 on NCBI Build GRCm38
Downstream adhesion G protein-coupled receptor F1 Adgrf1 5031409J19Rik,
Gpr110,
KPG_009
116.2kb Anti-sense Chr. 17 - 43270276 - 43325876 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002124.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045153.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045153.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045154.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045155.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 NR_045155.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019866.mm.1 TC1700002124.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0