This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277388_g1
See other CRACR2B GE Assays ›
Gene Symbol:
Cracr2b
Gene Name:
calcium release activated channel regulator 2B
Gene Aliases:
6330520A15Rik, BC026645, Cracr2a, Efcab4, Efcab4a
Chromosome Location:
Chr.7: 141460874 - 141466613 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277388_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cracr2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025103.2 NP_001020274.2 9 - 10 1137 82
GenBank mRNA BC026645.1 - 6 - 7 711 82
BC034124.1 - 3 - 4 333 82
BC048916.1 - 9 - 10 1137 82
BC098333.1 - 9 - 10 1098 5256663 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002054.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cracr2b

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
cellular protein localization
regulation of store-operated calcium entry
calcium ion binding
protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589000 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589002 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588981 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588978 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589003 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588982 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588977 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC098333.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 NM_001025103.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC026645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC034124.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC048916.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC098333.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 NM_001025103.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC026645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC034124.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552879 BC048916.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751903 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861731 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545999 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7113 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50596 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59739 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114840 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2367843 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC098333.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 NM_001025103.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC026645.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC034124.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC048916.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC098333.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 NM_001025103.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC026645.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC034124.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552879 BC048916.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589017 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588983 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589011 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588990 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588981 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588967 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588975 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588996 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588979 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589004 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588994 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588963 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589000 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588984 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588991 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588997 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588972 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588986 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589003 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588970 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589001 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588974 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588992 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588977 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588987 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25588988 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25589016 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114840 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06396718 - -
Whole Mouse Genome G4122A A_52_P552879 BC098333.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 NM_001025103.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC026645.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC034124.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC048916.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC098333.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 NM_001025103.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC026645.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC034124.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P552879 BC048916.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117240 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114840 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112458 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112530 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112466 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112523 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112515 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112469 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112452 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588977 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588981 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589000 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589003 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588978 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25589002 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25588982 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588986 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589049 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588937 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588975 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589077 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588991 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588988 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588997 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25588892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25589058 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588965 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589001 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589000 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06539135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588970 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06408312 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588990 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589012 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06422455 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588972 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06422451 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589017 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06112468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589015 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06539152 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06539155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06112523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588986 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588975 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588996 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588994 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588974 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589016 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06408319 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588967 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589006 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588979 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06422453 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589005 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25589007 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25588988 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10558937 - -
Affymetrix Mouse Gene 1.0 ST Array 10558940 - -
Affymetrix Mouse Gene 1.0 ST Array 10558945 - -
Affymetrix Mouse Gene 1.0 ST Array 10558947 - -
Affymetrix Mouse Gene 1.0 ST Array 10558944 - -
Affymetrix Mouse Gene 1.0 ST Array 10558941 - -
Affymetrix Mouse Gene 1.0 ST Array 10558943 - -
Affymetrix Mouse Gene 1.0 ST Array 10558946 - -
Affymetrix Mouse Gene 1.0 ST Array 10558938 - -
Affymetrix Mouse Gene 1.0 ST Array 10558942 - -
Affymetrix Mouse Gene 1.0 ST Array 10558939 - -
Affymetrix HT MG-430 PM Array Plate 1427367_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10558936 - -
Mouse Genome 430 2.0 Array 1427367_at - -
Mouse Genome 430 2.0 Array 1427367_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 213573_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 213573_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700018291.mm.1 NM_001025103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700018291.mm.1 NM_001025103.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700018291.mm.1 TC0700002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700018292.mm.1 NM_001025103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700018292.mm.1 NM_001025103.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700018292.mm.1 TC0700002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700018290.mm.1 NM_001025103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700018290.mm.1 NM_001025103.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700018290.mm.1 TC0700002054.mm.1 MTA TC
Mouse Expression Set 430 1427367_PM_at - -
Mouse Expression Set 430 1427367_at - -
Mouse Genome 430A 2.0 Array 1427367_PM_at - -
Mouse Genome 430A 2.0 Array 1427367_at - -
Affymetrix HT Mouse Genome 430A Array 1427367_at - -
Affymetrix HT Mouse Genome 430A Array 1427367_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1238203 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2720663 - -
MouseRef-8 v2 Expression BeadChip ILMN_2913222 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913222 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0