This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277386_gH
See other CRACR2B GE Assays ›
Gene Symbol:
Cracr2b
Gene Name:
calcium release activated channel regulator 2B
Gene Aliases:
6330520A15Rik, BC026645, Cracr2a, Efcab4, Efcab4a
Chromosome Location:
Chr.7: 141460874 - 141466613 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277386_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cracr2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025103.2 NP_001020274.2 7 - 8 889 92
GenBank mRNA BC026645.1 - 4 - 5 463 92
BC034124.1 - 1 - 2 85 92
BC048916.1 - 7 - 8 889 92
BC098333.1 - 7 - 8 850 5256663 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002054.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cracr2b

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
cellular protein localization
regulation of store-operated calcium entry
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700018285.mm.1 NM_001025103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700018285.mm.1 NM_001025103.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700018285.mm.1 TC0700002054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0