Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200012216.mm.1 |
NM_001003915.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012216.mm.1 |
NM_001003915.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012216.mm.1 |
NM_001177624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012216.mm.1 |
NM_001177624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012216.mm.1 |
TC0200001634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012217.mm.1 |
NM_001003915.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012217.mm.1 |
NM_001003915.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012217.mm.1 |
NM_001177624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012217.mm.1 |
NM_001177624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012217.mm.1 |
TC0200001634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012215.mm.1 |
NM_001003915.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012215.mm.1 |
NM_001003915.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012215.mm.1 |
NM_001177624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012215.mm.1 |
NM_001177624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012215.mm.1 |
TC0200001634.mm.1 |
MTA TC |