This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276425_m1
See other INPP5A GE Assays ›
Gene Symbol:
Inpp5a
Gene Name:
inositol polyphosphate-5-phosphatase A
Gene Aliases:
-
Chromosome Location:
Chr.7: 139389109 - 139579652 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276425_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127363.1 NP_001120835.1 9 - 10 1013 63
NM_183144.3 NP_898967.2 9 - 10 1013 63
GenBank mRNA AK083449.1 - 10 - 11 1012 63
AK157558.1 - 9 - 10 876 63
AK171360.1 - 9 - 10 1013 63
AK171880.1 - 9 - 10 989 63
BC056341.1 - 9 - 10 1013 5703345 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001991.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp5a

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

inositol phosphate dephosphorylation
inositol-polyphosphate 5-phosphatase activity
PH domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017476.mm.1 NM_001127363.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017476.mm.1 NM_001127363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017476.mm.1 NM_183144.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017476.mm.1 NM_183144.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700017476.mm.1 TC0700001991.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0