This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276321_m1
See other NSMCE4A GE Assays ›
Gene Symbol:
Nsmce4a
Gene Name:
non-SMC element 4 homolog A (S. cerevisiae)
Gene Aliases:
2410003A14Rik
Chromosome Location:
Chr.7: 130532526 - 130547381 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nsmce4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162855.1 NP_001156327.1 6 - 7 942 147
GenBank mRNA AK010349.1 - 6 - 7 942 147
AK010795.1 - 6 - 7 908 147
AK189457.1 - 2 - 3 139 147
BC032990.1 - 5 - 6 147
BC051572.1 - 5 - 6 147
BC089629.1 - 3 - 4 300 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004388.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of response to DNA damage stimulus
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700036953.mm.1 NM_001162855.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036953.mm.1 NM_001162855.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036953.mm.1 TC0700004388.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0