This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275981_g1
See other XRN2 GE Assays ›
Gene Symbol:
Xrn2
Gene Name:
5'-3' exoribonuclease 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 147012631 - 147078002 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275981_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xrn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011917.2 NP_036047.2 8 - 9 755 82
GenBank mRNA AK004800.1 - 8 - 9 730 82
AK011711.1 - 8 - 9 753 82
AK031247.1 - 8 - 9 798 82
AK053643.1 - 8 - 9 748 82
AK133654.1 - 8 - 9 737 82
AK163899.1 - 7 - 8 808 82
AK167019.1 - 8 - 9 775 82
AK168037.1 - 8 - 9 801 82
BC054743.1 - 8 - 9 755 6400974 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002178.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xrn2

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

DNA catabolic process, exonucleolytic
nucleobase-containing compound metabolic process
DNA repair
DNA recombination
transcription, DNA-templated
DNA-templated transcription, termination
regulation of transcription, DNA-templated
mRNA processing
microtubule-based process
cell growth
RNA metabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription termination site sequence-specific DNA binding
nucleic acid binding
DNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
protein binding
zinc ion binding
5'-3' exonuclease activity
hydrolase activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200017267.mm.1 NM_011917.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017267.mm.1 NM_011917.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200017267.mm.1 TC0200002178.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017266.mm.1 NM_011917.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017266.mm.1 NM_011917.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200017266.mm.1 TC0200002178.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0