This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275906_g1
See other CTTN GE Assays ›
Gene Symbol:
Cttn
Gene Name:
cortactin
Gene Aliases:
1110020L01Rik, Ems1
Chromosome Location:
Chr.7: 144435723 - 144471004 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275906_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cttn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252572.1 NP_001239501.1 13 - 14 1407 124
NM_007803.5 NP_031829.2 14 - 15 1518 124
GenBank mRNA AK084249.1 - 13 - 14 1217 124
AK146856.1 - 14 - 15 1325 124
AK148010.1 - 14 - 15 1321 124
AK165555.1 - 13 - 14 1210 124
AK168699.1 - 14 - 15 1319 124
AK168871.1 - 13 - 14 1208 124
BC011434.1 - 13 - 14 1200 3981559 124
BC117820.1 - 13 - 14 1220 124
U03184.1 - 14 - 15 1307 124

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cttn

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
endocytosis
receptor-mediated endocytosis
substrate-dependent cell migration, cell extension
actin filament polymerization
regulation of axon extension
positive regulation of actin filament polymerization
actin cytoskeleton reorganization
positive regulation of smooth muscle contraction
focal adhesion assembly
neuron projection morphogenesis
cell motility
dendritic spine maintenance
lamellipodium organization
regulation of mitophagy
negative regulation of extrinsic apoptotic signaling pathway
protein binding
profilin binding
proline-rich region binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700038954.mm.1 NM_001252572.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038954.mm.1 NM_001252572.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038954.mm.1 NM_007803.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038954.mm.1 NM_007803.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700038954.mm.1 TC0700004604.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0