This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275819_m1
See other SLC12A1 GE Assays ›
Gene Symbol:
Slc12a1
Gene Name:
solute carrier family 12, member 1
Gene Aliases:
AI788571, D630042G03Rik, Nkcc2, mBSC1, urehr3
Chromosome Location:
Chr.2: 125152600 - 125230002 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275819_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc12a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079690.1 NP_001073158.1 17 - 18 2369 81
NM_183354.2 NP_899197.2 17 - 18 2369 81
GenBank mRNA BC016888.1 - 16 - 17 2347 4236020 81
U20973.1 - 17 - 18 2367 81
U20974.1 - 17 - 18 2367 81
U20975.1 - 17 - 18 2367 81
U94518.1 - 16 - 17 2316 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001879.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc12a1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

kidney development
transport
ion transport
potassium ion transport
sodium ion transport
chloride transport
excretion
protein insertion into membrane from inner side
ion transmembrane transport
renal rubidium ion absorption
chemical homeostasis
transmembrane transport
positive regulation of calcineurin-NFAT signaling cascade
cellular hyperosmotic salinity response
cation transmembrane transport
chloride transmembrane transport
transporter activity
protein binding
sodium:potassium:chloride symporter activity
symporter activity
cation:chloride symporter activity
rubidium ion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200014640.mm.1 NM_001079690.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014640.mm.1 NM_001079690.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014640.mm.1 NM_183354.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014640.mm.1 NM_183354.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200014640.mm.1 TC0200001879.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0