This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275694_g1
See other EPS8L2 GE Assays ›
Gene Symbol:
Eps8l2
Gene Name:
EPS8-like 2
Gene Aliases:
AI042819, AW545405, Eps8r2
Chromosome Location:
Chr.7: 141338911 - 141363016 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275694_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eps8l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133191.2 NP_573454.2 13 - 14 1313 95
GenBank mRNA BC005492.1 - 13 - 14 1313 3487337 95
BC009098.1 - 12 - 13 1597 3482190 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002047.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eps8l2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
Rac protein signal transduction
regulation of Rho protein signal transduction
positive regulation of GTPase activity
positive regulation of ruffle assembly
actin binding
Rho guanyl-nucleotide exchange factor activity
Rac guanyl-nucleotide exchange factor activity
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700018176.mm.1 NM_133191.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700018176.mm.1 NM_133191.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700018176.mm.1 TC0700002047.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0