This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275580_m1
See other TGFB1I1 GE Assays ›
Gene Symbol:
Tgfb1i1
Gene Name:
transforming growth factor beta 1 induced transcript 1
Gene Aliases:
ARA55, Hic5, TSC-5, hic-5
Chromosome Location:
Chr.7: 128246799 - 128255699 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275580_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgfb1i1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289553.1 NP_001276482.1 1 - 2 198 58
GenBank mRNA AK085042.1 - 1 - 2 40 58
DQ143891.1 - 1 - 2 199 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001289553.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

response to heat
positive regulation of epithelial to mesenchymal transition
Wnt signaling pathway
morphogenesis of embryonic epithelium
cell differentiation
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
ubiquitin-dependent SMAD protein catabolic process
epithelial cell differentiation
cell fate commitment
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
transcription coactivator activity
protein binding
zinc ion binding
metal ion binding
Roundabout binding
androgen receptor binding
I-SMAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700016385.mm.1 NM_001289550.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016385.mm.1 NM_001289551.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016385.mm.1 NM_001289552.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016385.mm.1 NM_001289553.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016385.mm.1 TC0700001879.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700016375.mm.1 NM_001289550.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016375.mm.1 NM_001289551.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016375.mm.1 NM_001289552.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016375.mm.1 NM_001289553.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016375.mm.1 TC0700001879.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700016376.mm.1 NM_001289550.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016376.mm.1 NM_001289551.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016376.mm.1 NM_001289552.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016376.mm.1 NM_001289553.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016376.mm.1 TC0700001879.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700016379.mm.1 NM_001289550.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016379.mm.1 NM_001289551.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016379.mm.1 NM_001289552.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016379.mm.1 NM_001289553.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016379.mm.1 TC0700001879.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0