Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700016727.mm.1 |
NM_019564.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016727.mm.1 |
NM_019564.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016727.mm.1 |
TC0700001913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016728.mm.1 |
NM_019564.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016728.mm.1 |
NM_019564.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016728.mm.1 |
TC0700001913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016730.mm.1 |
NM_019564.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016730.mm.1 |
NM_019564.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016730.mm.1 |
TC0700001913.mm.1 |
MTA TC |