This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275445_m1
See other DUOXA1 GE Assays ›
Gene Symbol:
Duoxa1
Gene Name:
dual oxidase maturation factor 1
Gene Aliases:
BC019755, Nip, Nip1
Chromosome Location:
Chr.2: 122303540 - 122313744 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275445_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Duoxa1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC019755.1 - 3 - 4 186 3964798 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC019755.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

oxidase oxidoreductase metabolite interconversion enzyme

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200038316.mm.1 TC0200004507.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200038315.mm.1 TC0200004507.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 486381 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0