This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275042_m1
See other GUCA1B GE Assays ›
Gene Symbol:
Guca1b Actr1b
Gene Name:
guanylate cyclase activator 1B
ARP1 actin-related protein 1B, centractin beta
Gene Aliases:
BC018258, GCAP2
2310066K23Rik, AA960180, AI851923, Arp1b
Chromosome Location:
Chr.17: 47384694 - 47392967 on Build GRCm38
Chromosome Location:
Chr.1: 36698114 - 36710017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm01275042_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Guca1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146079.1 NP_666191.1 2 - 3 452 65
GenBank mRNA AK033052.1 - 2 - 3 417 65
BC018258.1 - 2 - 3 380 4511650 65
BC018480.1 - 2 - 3 421 4500919 65
BC029017.1 - 2 - 3 397 5359531 65
BC029661.1 - 2 - 3 407 5366068 65
BC031367.1 - 2 - 3 415 5358667 65
BC031916.1 - 2 - 3 452 5369642 65
BC033348.1 - 2 - 3 402 65
BC033358.1 - 2 - 3 451 5368968 65
BC033364.1 - 2 - 3 402 65
BC033380.1 - 2 - 3 407 5366953 65
BC033383.1 - 2 - 3 408 5368064 65
BC033396.1 - 2 - 3 399 65
BC033423.1 - 2 - 3 404 65
BC034331.1 - 2 - 3 408 5357797 65
BC034595.1 - 2 - 3 407 5362191 65
Gene Symbol: Actr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC027134.1 - 371 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000913.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Guca1b
Click here to find siRNAs for Gene Actr1b

Gene Ontology Categories:

Function(s) Process(es)

visual perception
phototransduction
regulation of guanylate cyclase activity
positive regulation of guanylate cyclase activity
response to stimulus
calcium ion binding
calcium sensitive guanylate cyclase activator activity
guanylate cyclase regulator activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700008751.mm.1 NM_146079.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008751.mm.1 NM_146079.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008751.mm.1 TC1700000913.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0