This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273903_g1
See other STAP2 GE Assays ›
Gene Symbol:
Stap2
Gene Name:
signal transducing adaptor family member 2
Gene Aliases:
AW049765, STAP-2
Chromosome Location:
Chr.17: 55997074 - 56005643 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273903_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145934.2 NP_666046.1 10 - 11 1082 73
NR_102313.1 9 - 10 1186 73
GenBank mRNA AK050131.1 - 10 - 11 1062 73
AK050172.1 - 9 - 10 1154 73
BC026642.1 - 10 - 11 1056 4986113 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002308.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stap2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700021529.mm.1 NM_145934.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700021529.mm.1 NR_102313.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700021529.mm.1 TC1700002308.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0