This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273877_m1
See other METAP1D GE Assays ›
Gene Symbol:
Metap1d
Gene Name:
methionyl aminopeptidase type 1D (mitochondrial)
Gene Aliases:
2310066F24Rik, 3110033D18Rik, AV117938, MAP 1D, Map1d, MetAP 1D, Metapl1
Chromosome Location:
Chr.2: 71453319 - 71525196 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273877_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Metap1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025633.3 NP_079909.1 7 - 8 819 82
GenBank mRNA AJ414378.1 - 4 - 5 682 82
AK005241.1 - 7 - 8 816 82
AK010067.1 - 7 - 8 819 82
BC051534.1 - 4 - 5 693 3466745 82
BC060739.1 - 6 - 7 2327 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001038.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Metap1d

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
peptidase activity
metalloexopeptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200008396.mm.1 NM_025633.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008396.mm.1 NM_025633.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200008396.mm.1 TC0200001038.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200008397.mm.1 NM_025633.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008397.mm.1 NM_025633.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200008397.mm.1 TC0200001038.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0