This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273660_g1
See other MEA1 GE Assays ›
Gene Symbol:
Mea1
Gene Name:
male enhanced antigen 1
Gene Aliases:
Mea-1
Chromosome Location:
Chr.17: 46679138 - 46683126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273660_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mea1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102392.1 1 - 2 181 77
GenBank mRNA AK005106.1 - 1 - 2 38 77
AK005369.1 - 1 - 2 42 77
AK008194.1 - 1 - 2 48 77
AK008305.1 - 1 - 2 50 77
AK010863.1 - 1 - 2 59 77
AK010962.1 - 1 - 2 44 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005106.1,AK005369.1,AK008194.1,AK008305.1,AK010863.1,AK010962.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0