This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273461_g1
See other GUCY2D GE Assays ›
Gene Symbol:
Gucy2d
Gene Name:
guanylate cyclase 2d
Gene Aliases:
-
Chromosome Location:
Chr.7: 98440416 - 98477572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273461_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130693.3 NP_001124165.1 5 - 6 1600 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001366.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
guanylate cyclase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700011626.mm.1 NM_001130693.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700011626.mm.1 TC0700001366.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0