This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01272663_m1
See other METTL5OS GE Assays ›
Gene Symbol:
Mettl5os
Gene Name:
methyltransferase like 5, opposite strand
Gene Aliases:
4930550G17Rik
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01272663_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl5os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016085.1 - 1 - 2 378 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52402 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298872 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151335 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151335 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151335 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432524_PM_at - -
Mouse Genome 430 2.0 Array 1432524_PM_at - -
Mouse Genome 430 2.0 Array 1432524_at - -
Mouse Expression Set 430 1432524_PM_at - -
Mouse Expression Set 430 1432524_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 336014 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4780070 - -
Illumina mouse-6 v1.0 expression beadchip 105080152 - -
Illumina Sentrix mouse-6.v1 bead array 105080152 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105080152 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2572840 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0