This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01271642_m1
See other PDE3B GE Assays ›
Gene Symbol:
Pde3b
Gene Name:
phosphodiesterase 3B, cGMP-inhibited
Gene Aliases:
9830102A01Rik, AI847709
Chromosome Location:
Chr.7: 114414700 - 114537938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01271642_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011055.2 NP_035185.2 14 - 15 3108 64
GenBank mRNA AJ132271.1 - 14 - 15 2811 64
X95521.1 - 14 - 15 1953 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001669.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde3b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
negative regulation of angiogenesis
endocrine pancreas development
glucose homeostasis
negative regulation of insulin secretion
regulation of insulin secretion
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
drug binding
hydrolase activity
cAMP binding
protein kinase B binding
metal ion binding
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700013921.mm.1 NM_011055.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700013921.mm.1 NM_011055.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700013921.mm.1 TC0700001669.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0