This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270050_m1
See other EDRF1 GE Assays ›
Gene Symbol:
Edrf1
Gene Name:
erythroid differentiation regulatory factor 1
Gene Aliases:
2700050L05Rik, AU022667, AW558805
Chromosome Location:
Chr.7: 133637536 - 133673017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270050_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Edrf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178115.4 NP_835216.3 17 - 18 2510 128
GenBank mRNA AK144179.1 - 17 - 18 2469 128
BC024916.1 - 5 - 6 642 128
BC046235.1 - 5 - 6 860 6817045 128
BC072596.1 - 17 - 18 2479 6409214 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001947.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Edrf1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017049.mm.1 NM_178115.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017049.mm.1 NM_178115.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700017049.mm.1 TC0700001947.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700017050.mm.1 NM_178115.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017050.mm.1 NM_178115.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700017050.mm.1 TC0700001947.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0