This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01269624_g1
See other DKK3 GE Assays ›
Gene Symbol:
Dkk3
Gene Name:
dickkopf homolog 3 (Xenopus laevis)
Gene Aliases:
AW061014, C87148
Chromosome Location:
Chr.7: 112116019 - 112159121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01269624_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dkk3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015814.2 NP_056629.1 1 - 2 95 64
GenBank mRNA AF177400.1 - 1 - 2 82 64
AJ243964.1 - 1 - 2 98 64
AK004853.1 - 1 - 2 95 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF177400.1, AJ243964.1, AK004853.1, NM_015814.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dkk3

Gene Ontology Categories:

Function(s)

multicellular organism development
Wnt signaling pathway
negative regulation of Wnt signaling pathway
negative regulation of aldosterone biosynthetic process
negative regulation of transcription, DNA-templated
negative regulation of canonical Wnt signaling pathway
negative regulation of cortisol biosynthetic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0