This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01269445_m1
See other ADGRE4 GE Assays ›
Gene Symbol:
Adgre4
Gene Name:
adhesion G protein-coupled receptor E4
Gene Aliases:
D17Ertd479e, Egf-tm7, Emr4, Fire, Gpr127
Chromosome Location:
Chr.17: 55749984 - 55853662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01269445_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgre4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139138.3 NP_631877.2 7 - 8 701 121
GenBank mRNA AF396935.1 - 7 - 8 659 121
AK037483.1 - 7 - 8 646 121
AK155935.1 - 7 - 8 649 121
AY032690.1 - 7 - 8 620 121
BC120538.1 - 7 - 8 658 40129461 121
BC120540.1 - 7 - 8 658 40129463 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000990.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgre4

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
epidermal growth factor receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009414.mm.1 NM_139138.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009414.mm.1 NM_139138.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009414.mm.1 TC1700000990.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0