This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01269297_m1
See other MUT GE Assays ›
Gene Symbol:
Mut
Gene Name:
methylmalonyl-Coenzyme A mutase
Gene Aliases:
D230010K02Rik, Mcm
Chromosome Location:
Chr.17: 40934685 - 40961989 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01269297_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mut
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008650.3 NP_032676.2 8 - 9 1692 71
GenBank mRNA AK146309.1 - 8 - 9 1652 71
BC019175.1 - 8 - 9 1674 5007694 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000819.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mut

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
post-embryonic development
homocysteine metabolic process
catalytic activity
methylmalonyl-CoA mutase activity
isomerase activity
intramolecular transferase activity
cobalamin binding
metal ion binding
modified amino acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700007932.mm.1 NM_008650.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700007932.mm.1 NM_008650.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700007932.mm.1 TC1700000819.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm