This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01269183_mH
See other CLIC5 GE Assays ›
Gene Symbol:
Clic5
Gene Name:
chloride intracellular channel 5
Gene Aliases:
5730531E12Rik, B330005L24, Gm322, jbg
Chromosome Location:
Chr.17: 44134678 - 44280168 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01269183_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clic5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172621.2 NP_766209.1 2 - 3 503 58
GenBank mRNA AK040195.1 - 1 - 2 141 58
AK046522.1 - 2 - 3 492 58
AK156849.1 - 2 - 3 344 58
BC064037.1 - 2 - 3 488 1227040 58

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clic5

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to dietary excess
diet induced thermogenesis
transport
ion transport
chloride transport
sensory perception of sound
protein localization
regulation of ion transmembrane transport
neuromuscular process controlling balance
auditory receptor cell stereocilium organization
chloride transmembrane transport
voltage-gated ion channel activity
chloride channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0