This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268643_g1
See other STAM2 GE Assays ›
Gene Symbol:
Stam2
Gene Name:
signal transducing adaptor molecule (SH3 domain and ITAM motif) 2
Gene Aliases:
1200004O12Rik, 5730456G07Rik, BB145015, Hbp
Chromosome Location:
Chr.2: 52692198 - 52742149 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268643_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stam2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019667.2 NP_062641.1 9 - 10 938 146
GenBank mRNA AB012611.1 - 9 - 10 953 146
AK004604.2 - 9 - 10 938 146
AK035789.1 - 991 146
AK044230.1 - 8 - 9 867 146
AK159699.1 - 9 - 10 967 146
AK168729.1 - 9 - 10 970 146
BC013818.1 - 9 - 10 974 3671426 146

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003540.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stam2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200030244.mm.1 NM_019667.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030244.mm.1 NM_019667.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200030244.mm.1 TC0200003540.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0