This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266695_g1
See other TTC39AOS1 GE Assays ›
Gene Symbol:
Ttc39aos1
Gene Name:
Ttc39a opposite strand RNA 1
Gene Aliases:
Gm12750, OTTMUSG00000008290, lincRNA-EPS
Chromosome Location:
Chr.4: 109402273 - 109406267 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266695_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttc39aos1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK148461.1 - 1 - 2 1736 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tetratricopeptide repeat domain 39A Ttc39a 4922503N01Rik Anti-sense Chr. 4 - 109394148 - 109444745 on NCBI Build GRCm38
Downstream ring finger protein 11 Rnf11 46.2kb Sense Chr. 4 - 109452473 - 109484723 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0