Assay Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008891.2 | NP_032917.2 | 5 - 6 | 503 | 80 | |
| GenBank mRNA | AK009055.1 | - | 5 - 6 | 458 | 80 | |
| AK028219.1 | - | 5 - 6 | 444 | 80 | ||
| AK030978.1 | - | 5 - 6 | 432 | 80 | ||
| AK050669.1 | - | 5 - 6 | 449 | 80 | ||
| AK051640.1 | - | 5 - 6 | 501 | 80 | ||
| AK157587.1 | - | 5 - 6 | 447 | 80 | ||
| AK160616.1 | - | 5 - 6 | 438 | 80 | ||
| AK161109.1 | - | 5 - 6 | 443 | 80 | ||
| BC132571.1 | - | 5 - 6 | 437 | 40130848 | 80 | |
| BC138014.1 | - | 5 - 6 | 437 | 8861033 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnn
Genomic Map