This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265810_m1
See other GPRC5D GE Assays ›
Gene Symbol:
Gprc5d
Gene Name:
G protein-coupled receptor, family C, group 5, member D
Gene Aliases:
-
Chromosome Location:
Chr.6: 135105991 - 135137443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265810_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gprc5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205396.1 NP_001192325.1 2 - 3 1046 71
GenBank mRNA AB025221.1 - 1 - 2 892 71
AK137739.1 - 2 - 3 1009 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
keratinization
hair cycle
molecular_function
signal transducer activity
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0