This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265807_m1
See other GPRC5A GE Assays ›
Gene Symbol:
Gprc5a
Gene Name:
G protein-coupled receptor, family C, group 5, member A
Gene Aliases:
Rai3, Raig1
Chromosome Location:
Chr.6: 135065662 - 135084709 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265807_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gprc5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181444.5 NP_852109.2 1 - 2 121 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_181444.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gprc5a

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600012830.mm.1 NM_181444.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600012830.mm.1 NM_181444.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600012830.mm.1 TC0600001602.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600012829.mm.1 NM_181444.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600012829.mm.1 NM_181444.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600012829.mm.1 TC0600001602.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0