This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265205_m1
See other LEPROT GE Assays ›
Gene Symbol:
Leprot
Gene Name:
leptin receptor overlapping transcript
Gene Aliases:
LEPR, OB-RGRP, Obrgrp, VPS55
Chromosome Location:
Chr.4: 101647783 - 101659358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265205_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Leprot
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175036.4 NP_778201.1 1 - 2 46 66
GenBank mRNA AK009569.1 - 1 - 2 47 66
AK152008.1 - 1 - 2 113 66
AK159489.1 - 1 - 2 112 66
AK160453.1 - 1 - 2 60 66
AK165821.1 - 1 - 2 105 66
AK168597.1 - 1 - 2 82 66
BC004744.1 - 1 - 2 57 3583760 66
BC010289.1 - 1 - 2 36 2651399 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK009569.1, AK159489.1, AK160453.1, AK165821.1, AK168597.1, BC004744.1, BC010289.1, NM_175036.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Leprot

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of JAK-STAT cascade
negative regulation of growth hormone receptor signaling pathway
positive regulation of protein targeting to mitochondrion
negative regulation of protein localization to cell surface
receptor binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400007049.mm.1 NM_175036.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007049.mm.1 NM_175036.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400007049.mm.1 TC0400001038.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400007048.mm.1 NM_175036.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007048.mm.1 NM_175036.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400007048.mm.1 TC0400001038.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 900230 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0