This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265100_m1
See other RBBP8 GE Assays ›
Gene Symbol:
Rbbp8
Gene Name:
retinoblastoma binding protein 8
Gene Aliases:
9930104E21Rik, CtIP, RBBP-8, RIM, SAE2
Chromosome Location:
Chr.18: 11633276 - 11743207 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265100_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbbp8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081223.2 NP_001074692.1 10 - 11 1357 74
NM_001252495.1 NP_001239424.1 10 - 11 1262 74
NR_045526.1 10 - 11 1357 74
NR_045527.1 9 - 10 1201 74
GenBank mRNA AK037049.1 - 10 - 11 1280 74
AK041384.1 - 9 - 10 1107 74
AK077614.1 - 9 - 10 1309 74
BC019665.1 - 7 - 8 702 74
BC027367.1 - 10 - 11 1225 74
BC050849.1 - 10 - 11 1262 6508067 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000092.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
double-strand break repair via homologous recombination
blastocyst hatching
DNA repair
nucleotide-excision repair
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
cell cycle
mitotic nuclear division
DNA double-strand break processing involved in repair via single-strand annealing
G2 DNA damage checkpoint
cell division
meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
RNA polymerase II repressing transcription factor binding
RNA polymerase II transcription corepressor activity
DNA binding
damaged DNA binding
nuclease activity
endonuclease activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NM_001081223.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NM_001081223.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NM_001252495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NM_001252495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NR_045526.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NR_045526.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NR_045527.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 NR_045527.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000617.mm.1 TC1800000092.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0