Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900006708.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006708.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006708.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006712.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006712.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006712.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006718.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006718.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006718.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006719.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006719.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006719.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006715.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006715.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006715.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006711.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006711.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006711.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006713.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006713.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006713.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006714.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006714.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006714.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006721.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006721.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006721.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006710.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006710.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006710.mm.1 |
TC1900000760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006717.mm.1 |
NM_001170847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006717.mm.1 |
NM_001170847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006717.mm.1 |
TC1900000760.mm.1 |
MTA TC |