This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264214_m1
See other NKX2-1 GE Assays ›
Gene Symbol:
Nkx2-1
Gene Name:
NK2 homeobox 1
Gene Aliases:
AV026640, Nkx2.1, T/EBP, Titf1, Ttf-1
Chromosome Location:
Chr.12: 56531935 - 56536908 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264214_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nkx2-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009385.3 NP_033411.3 1 - 2 604 99
GenBank mRNA BC057607.1 - 1 - 2 604 6416507 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC057607.1, NM_009385.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nkx2-1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neuron migration
regulation of blood volume by renin-angiotensin
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
phospholipid metabolic process
pattern specification process
axon guidance
brain development
endoderm development
locomotory behavior
feeding behavior
organ morphogenesis
positive regulation of gene expression
negative regulation of epithelial to mesenchymal transition
telencephalon development
globus pallidus development
hippocampus development
cerebral cortex cell migration
forebrain dorsal/ventral pattern formation
forebrain neuron fate commitment
forebrain neuron differentiation
cerebral cortex GABAergic interneuron differentiation
cerebral cortex neuron differentiation
pituitary gland development
telencephalon cell migration
lung development
negative regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
thyroid gland development
developmental induction
response to lipopolysaccharide
Leydig cell differentiation
hyperosmotic salinity response
menarche
positive regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
anatomical structure formation involved in morphogenesis
neuron fate commitment
oligodendrocyte differentiation
lung saccule development
Clara cell differentiation
Type II pneumocyte differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region DNA binding
core promoter binding
intronic transcription regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
enzyme binding
sequence-specific DNA binding
transcription regulatory region DNA binding
intronic transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200012314.mm.1 NM_001146198.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012314.mm.1 NM_001146198.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012314.mm.1 NM_009385.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012314.mm.1 NM_009385.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200012314.mm.1 TC1200001791.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200012316.mm.1 NM_009385.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012316.mm.1 NM_009385.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200012316.mm.1 TC1200001791.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0