This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264125_g1
See other ZYG11B GE Assays ›
Gene Symbol:
Zyg11b
Gene Name:
zyg-ll family member B, cell cycle regulator
Gene Aliases:
1110046I03Rik, 2810482G21Rik, D4Mgi23, Gm431, mKIAA1730
Chromosome Location:
Chr.4: 108227755 - 108301141 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264125_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zyg11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033634.3 NP_001028806.2 7 - 8 1603 61
GenBank mRNA AK148335.1 - 7 - 8 1602 61
AK148760.1 - 1 - 2 73 61
AK166031.1 - 7 - 8 1583 61
BC139383.1 - 7 - 8 1570 8862405 61
BC139384.1 - 7 - 8 1570 8862406 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of ubiquitin-protein transferase activity
molecular_function
ubiquitin-protein transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0