This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264123_m1
See other ZYG11B GE Assays ›
Gene Symbol:
Zyg11b
Gene Name:
zyg-ll family member B, cell cycle regulator
Gene Aliases:
1110046I03Rik, 2810482G21Rik, D4Mgi23, Gm431, mKIAA1730
Chromosome Location:
Chr.4: 108227755 - 108301141 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264123_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zyg11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033634.3 NP_001028806.2 5 - 6 1439 82
GenBank mRNA AK145130.1 - 5 - 6 1412 82
AK148335.1 - 5 - 6 1438 82
AK162646.1 - 5 - 6 1402 82
AK166031.1 - 5 - 6 1419 82
AK166174.1 - 5 - 6 1419 82
AK166278.1 - 5 - 6 1429 82
BC117977.2 - 5 - 6 1403 40099428 82
BC117978.2 - 5 - 6 1403 40099429 82
BC139383.1 - 5 - 6 1406 8862405 82
BC139384.1 - 5 - 6 1406 8862406 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003237.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of ubiquitin-protein transferase activity
molecular_function
ubiquitin-protein transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400025798.mm.1 NM_001033634.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025798.mm.1 NM_001033634.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400025798.mm.1 TC0400003237.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0