This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264039_m1
See other HDHD2 GE Assays ›
Gene Symbol:
Hdhd2
Gene Name:
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases:
0610039H12Rik, 3110052N05Rik
Chromosome Location:
Chr.18: 76944115 - 76972171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264039_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdhd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039202.1 NP_001034291.1 4 - 5 480 82
GenBank mRNA AK151285.1 - 4 - 5 468 82
AK152162.1 - 4 - 5 468 82
AK152259.1 - 4 - 5 468 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001746.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800005964.mm.1 NM_001039202.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005964.mm.1 NM_001039202.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005964.mm.1 TC1800001746.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0