This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264035_m1
See other HDHD2 GE Assays ›
Gene Symbol:
Hdhd2
Gene Name:
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases:
0610039H12Rik, 3110052N05Rik
Chromosome Location:
Chr.18: 76944115 - 76972171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264035_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdhd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039201.1 NP_001034290.1 5 - 6 736 56
NM_029826.2 NP_084102.1 5 - 6 697 56
GenBank mRNA AK002837.1 - 5 - 6 685 56
AK014208.1 - 5 - 6 691 56
AK137285.1 - 5 - 6 689 56
AK147794.1 - 718 56
BC058177.1 - 5 - 6 671 5250739 56

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdhd2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

enzyme binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0