This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264031_m1
See other HDHD2 GE Assays ›
Gene Symbol:
Hdhd2
Gene Name:
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases:
0610039H12Rik, 3110052N05Rik
Chromosome Location:
Chr.18: 76944115 - 76972171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264031_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdhd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039202.1 NP_001034291.1 1 - 2 71 93
NM_029826.2 NP_084102.1 1 - 2 71 93
GenBank mRNA AK002837.1 - 1 - 2 59 93
AK137285.1 - 1 - 2 63 93
AK151285.1 - 1 - 2 59 93
AK152162.1 - 1 - 2 59 93
AK152259.1 - 1 - 2 59 93
BC058177.1 - 1 - 2 45 5250739 93
BY069367.1 - 1 - 2 81 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK002837.1, AK137285.1, AK151285.1, AK152162.1, AK152259.1, BC058177.1, NM_001039202.1, NM_029826.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdhd2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

enzyme binding
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 572105 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0