Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10519227 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519230 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519229 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519231 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519232 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519233 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519225 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519228 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10511207 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519224 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0400034937.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034937.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034937.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034930.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034930.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034930.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034932.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034932.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034932.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034940.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034940.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034940.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034934.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034934.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034934.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034938.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034938.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034938.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034939.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034939.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034939.mm.1 |
TC0400004160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034931.mm.1 |
NM_001033490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034931.mm.1 |
NM_001033490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034931.mm.1 |
TC0400004160.mm.1 |
MTA TC |