This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01263703_m1
See other PGD GE Assays ›
Gene Symbol:
Pgd
Gene Name:
phosphogluconate dehydrogenase
Gene Aliases:
0610042A05Rik, AU019875, C78335
Chromosome Location:
Chr.4: 149149985 - 149166707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01263703_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081274.1 NP_001074743.1 4 - 5 444 86
GenBank mRNA AK002894.1 - 4 - 5 406 86
AK077110.1 - 4 - 5 406 86
AK145602.1 - 4 - 5 408 86
AK150210.1 - 4 - 5 407 86
AK150490.1 - 4 - 5 403 86
AK151380.1 - 4 - 5 403 86
AK151993.1 - 4 - 5 403 86
AK152758.1 - 4 - 5 403 86
AK152894.1 - 4 - 5 403 86
AK153409.1 - 4 - 5 400 86
AK155027.1 - 4 - 5 403 86
AK161869.1 - 4 - 5 402 86
AK166733.1 - 4 - 5 398 86
AK166947.1 - 4 - 5 380 86
AK167215.1 - 4 - 5 398 86
AK168251.1 - 4 - 5 403 86
BC008646.1 - 4 - 5 375 3155147 86
BC011329.1 - 4 - 5 393 3984654 86
BC014793.1 - 4 - 5 412 3155100 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
pentose-phosphate shunt
NADP metabolic process
pentose-phosphate shunt, oxidative branch
pentose biosynthetic process
D-gluconate metabolic process
oxidation-reduction process
phosphogluconate dehydrogenase (decarboxylating) activity
phosphogluconate 2-dehydrogenase activity
oxidoreductase activity
carbohydrate binding
carboxylic acid binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0