This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01263637_g1
See other TRP73 GE Assays ›
Gene Symbol:
Trp73
Gene Name:
transformation related protein 73
Gene Aliases:
TAp73, Tp73, deltaNp73, p73
Chromosome Location:
Chr.4: 154056249 - 154140208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01263637_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trp73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001126330.1 NP_001119802.1 6 - 7 1077 106
NM_001126331.1 NP_001119803.1 6 - 7 1077 106
NM_011642.3 NP_035772.2 8 - 9 1367 106
GenBank mRNA AB221594.1 - 7 - 8 959 106
AK014503.1 - 1 - 2 56 106
AK017412.1 - 5 - 6 608 106
AK139633.1 - 6 - 7 1078 106
BC066045.1 - 6 - 7 964 6812399 106
BC069182.1 - 2 - 3 224 106
Y19234.1 - 8 - 9 1368 106
Y19235.1 - 6 - 7 1071 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK014503.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trp73

Panther Classification:

Molecular Function -

P53-like transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
release of cytochrome c from mitochondria
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
inflammatory response
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
cell cycle
cell cycle arrest
regulation of mitotic cell cycle
post-embryonic development
response to X-ray
response to gamma radiation
regulation of gene expression
hippocampus development
forebrain development
mitotic G1 DNA damage checkpoint
cerebrospinal fluid secretion
cellular response to UV
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic process
negative regulation of JUN kinase activity
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of cell size
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract morphogenesis
neuron development
positive regulation of oligodendrocyte differentiation
protein tetramerization
negative regulation of cardiac muscle cell proliferation
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein kinase binding
identical protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_001126330.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_001126330.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_001126331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_001126331.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_011642.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 NM_011642.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400034350.mm.1 TC0400004116.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0