This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01263614_m1
See other FAF1 GE Assays ›
Gene Symbol:
Faf1
Gene Name:
Fas-associated factor 1
Gene Aliases:
AA408698, Dffrx, Fam
Chromosome Location:
Chr.4: 109676627 - 109963960 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01263614_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Faf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007983.2 NP_032009.2 10 - 11 1371 148
GenBank mRNA AK053752.1 - 10 - 11 1306 148
BC065098.1 - 10 - 11 1371 30362116 148
U39643.1 - 10 - 11 1381 148

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Faf1

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
cytoplasmic sequestering of NF-kappaB
cell death
positive regulation of cell death
regulation of cell adhesion
positive regulation of protein complex assembly
regulation of protein catabolic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
protein binding
protein kinase binding
protein domain specific binding
heat shock protein binding
ubiquitin protein ligase binding
ubiquitin binding
NF-kappaB binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0