This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262764_m1
See other CD74 GE Assays ›
Gene Symbol:
Cd74
Gene Name:
CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated)
Gene Aliases:
CLIP, DHLAG, HLADG, Ia-GAMMA, Ii
Chromosome Location:
Chr.18: 60803849 - 60812652 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262764_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042605.1 NP_001036070.1 4 - 5 472 63
NM_010545.3 NP_034675.1 4 - 5 472 63
GenBank mRNA AK002232.1 - 4 - 5 472 63
AK020607.1 - 4 - 5 473 63
AK154096.1 - 4 - 5 503 63
BC003476.1 - 4 - 5 472 2650401 63
BC096435.1 - 4 - 5 483 6530802 63
X00496.1 - 3 - 4 249 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000617.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd74

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
prostaglandin biosynthetic process
positive regulation of cytokine-mediated signaling pathway
immune system process
positive regulation of dendritic cell antigen processing and presentation
negative regulation of peptide secretion
positive regulation of adaptive immune response
positive regulation of type 2 immune response
negative regulation of mature B cell apoptotic process
protein folding
protein complex assembly
intracellular protein transport
defense response
immune response
signal transduction
cell proliferation
immunoglobulin mediated immune response
cytokine-mediated signaling pathway
antigen processing and presentation
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive regulation of B cell proliferation
macrophage migration inhibitory factor signaling pathway
negative regulation of apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive thymic T cell selection
negative thymic T cell selection
positive regulation of innate immune response
negative regulation of T cell differentiation
positive regulation of T cell differentiation
positive regulation of fibroblast proliferation
positive regulation of peptidyl-tyrosine phosphorylation
chaperone mediated protein folding requiring cofactor
positive regulation of macrophage cytokine production
positive regulation of ERK1 and ERK2 cascade
positive regulation of neutrophil chemotaxis
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of chemokine (C-X-C motif) ligand 2 production
beta-amyloid binding
cytokine receptor activity
protein binding
cytokine binding
MHC class II protein complex binding
macrophage migration inhibitory factor binding
MHC class II protein binding
MHC class II protein binding, via antigen binding groove
protein binding involved in protein folding
nitric-oxide synthase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800004411.mm.1 NM_001042605.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004411.mm.1 NM_001042605.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004411.mm.1 NM_010545.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004411.mm.1 NM_010545.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800004411.mm.1 TC1800000617.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0