This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262452_m1
See other ACAA2 GE Assays ›
Gene Symbol:
Acaa2
Gene Name:
acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase)
Gene Aliases:
0610011L04Rik, AI255831, AI265397, D18Ertd240e
Chromosome Location:
Chr.18: 74779212 - 74806207 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262452_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acaa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177470.3 NP_803421.1 3 - 4 349 77
GenBank mRNA AK002555.1 - 3 - 4 339 77
AK050101.1 - 3 - 4 351 77
AK146007.1 - 3 - 4 364 77
AK167567.1 - 3 - 4 350 77
AK167715.1 - 3 - 4 347 77
AK169359.1 - 3 - 4 350 77
BC028901.1 - 3 - 4 344 4195488 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acaa2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA metabolic process
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
cellular response to hypoxia
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
negative regulation of mitochondrial membrane permeability involved in apoptotic process
catalytic activity
acetyl-CoA C-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0