This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262283_m1
See other CCBE1 GE Assays ›
Gene Symbol:
Ccbe1
Gene Name:
collagen and calcium binding EGF domains 1
Gene Aliases:
4933426F18Rik, 9430093N24Rik, mKIAA1983
Chromosome Location:
Chr.18: 66056855 - 66291838 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262283_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccbe1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178793.4 NP_848908.1 4 - 5 481 61
GenBank mRNA AK028377.1 - 4 - 5 479 61
AK035153.1 - 4 - 5 476 61
AK039742.1 - 4 - 5 411 61
AK087673.1 - 499 61
AK220435.1 - 4 - 5 451 61
BC103803.1 - 4 - 5 431 30088062 61
BC152322.1 - 4 - 5 450 40140631 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001479.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccbe1

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
lymphangiogenesis
multicellular organism development
positive regulation of vascular endothelial growth factor production
positive regulation of endothelial cell migration
positive regulation of protein processing
positive regulation of angiogenesis
positive regulation of vascular endothelial growth factor signaling pathway
positive regulation of lymphangiogenesis
protease binding
calcium ion binding
collagen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800010929.mm.1 NM_178793.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010929.mm.1 NM_178793.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800010929.mm.1 TC1800001479.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800010930.mm.1 NM_178793.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010930.mm.1 NM_178793.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800010930.mm.1 TC1800001479.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0