This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262181_m1
See other PIP5K1A GE Assays ›
Gene Symbol:
Pip5k1a
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha
Gene Aliases:
PI4P5K-I[a], PIP5K1-alpha, PIP5KIalpha, Pipk5a
Chromosome Location:
Chr.3: 95058530 - 95106930 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262181_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pip5k1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293707.1 NP_001280636.1 2 - 3 598 87
NM_008847.3 NP_032873.2 2 - 3 595 87
GenBank mRNA AK098097.1 - 2 - 3 552 87
AK150898.1 - 2 - 3 555 87
AK151984.1 - 2 - 3 555 87
AK158062.1 - 2 - 3 540 87
BC003763.1 - 2 - 3 234 3591003 87
BC031774.1 - 2 - 3 523 4503697 87
D86177.1 - 2 - 3 385 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pip5k1a

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid biosynthetic process
fibroblast migration
phosphorylation
actin cytoskeleton reorganization
phosphatidylinositol metabolic process
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
nucleotide binding
protein binding
ATP binding
kinase activity
phosphatidylinositol phosphate kinase activity
1-phosphatidylinositol-4-phosphate 5-kinase activity
transferase activity
kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0