This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262174_g1
See other VPS72 GE Assays ›
Gene Symbol:
Vps72
Gene Name:
vacuolar protein sorting 72 (yeast)
Gene Aliases:
Tcfl1, YL-1
Chromosome Location:
Chr.3: 95111042 - 95123051 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262174_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vps72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009336.2 NP_033362.2 1 - 2 171 89
GenBank mRNA AK140284.1 - 1 - 2 169 89
AK155191.1 - 1 - 2 169 89
AK179387.1 - 1 - 2 70 89
BC004834.1 - 1 - 2 149 2651158 89
BC043029.1 - 1 - 2 172 5692301 89
BY294768.1 - 1 - 2 192 89
D43643.1 - 1 - 2 142 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vps72

Panther Classification:

Molecular Function -

DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
somatic stem cell population maintenance
DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0