This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261455_m1
See other PDE3A GE Assays ›
Gene Symbol:
Pde3a
Gene Name:
phosphodiesterase 3A, cGMP inhibited
Gene Aliases:
A930022O17Rik, C87899
Chromosome Location:
Chr.6: 141249269 - 141506863 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261455_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018779.1 NP_061249.1 4 - 5 1741 88
GenBank mRNA AF099187.1 - 4 - 5 1741 88
AK004638.1 - 4 - 5 1046 88
AK052374.1 - 4 - 5 1019 88
AK164864.1 - 4 - 5 522 88
BC138265.1 - 4 - 5 1699 8861284 88
BC138266.1 - 4 - 5 1699 8861285 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001664.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde3a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
cAMP catabolic process
signal transduction
cAMP-mediated signaling
cGMP-mediated signaling
regulation of meiotic nuclear division
response to drug
negative regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of vascular permeability
positive regulation of oocyte development
cellular response to cGMP
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
cAMP binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013275.mm.1 NM_018779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013275.mm.1 NM_018779.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013275.mm.1 TC0600001664.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0